Learn More
Medchemexpress LLC HY-N4262 5mg Medchemexpress, Ginsenoside Rh7 CAS:343780-68-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN42625MG
Encompass_Preferred
Medchemexpress, HY-N4262 5mg Ginsenoside Rh7 CAS:343780-68-7 Ginsenoside Rh7 is a minor saponin isolated from the leaves of Panax ginseng. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2931