Learn More
Medchemexpress LLC Ginsenoside Rk1 | 494753-69-4 | 20 MG
SDP
Catalog No. 5000420492
Encompass_Preferred
Ginsenoside Rk1, a compound derived from processed ginseng, exhibits a wide range of biological activities. It demonstrates anti-inflammatory, anti-tumor, and antimicrobial effects. It also suppresses the Jak2/Stat3 signaling pathway and NF-κB, while inducing cell apoptosis through ROS generation and blocking the PI3K/Akt pathway.
  • Anti-inflammatory effects
  • Suppresses Jak2/Stat3 signaling and NF-κB activation
  • Anti-tumor and antimicrobial properties
  • Antiplatelet aggregation activity
  • Anti-insulin resistance
  • Nephroprotective effects
  • Enhances cognitive function
  • Reduces lipid accumulation
  • Prevents osteoporosis
  • Induces cell apoptosis via ROS generation
  • Blocks PI3K/Akt pathway
Catalog No. 50-004-20492 Supplier Medchemexpress LLC Supplier No. HYN251520MG
Ginsenoside Rk1, a compound derived from processed ginseng, exhibits a wide range of biological activities. It demonstrates anti-inflammatory, anti-tumor, and antimicrobial effects. It also suppresses the Jak2/Stat3 signaling pathway and NF-κB, while inducing cell apoptosis through ROS generation and blocking the PI3K/Akt pathway.
  • Anti-inflammatory effects
  • Suppresses Jak2/Stat3 signaling and NF-κB activation
  • Anti-tumor and antimicrobial properties
  • Antiplatelet aggregation activity
  • Anti-insulin resistance
  • Nephroprotective effects
  • Enhances cognitive function
  • Reduces lipid accumulation
  • Prevents osteoporosis
  • Induces cell apoptosis via ROS generation
  • Blocks PI3K/Akt pathway