Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ginsenoside Rk1 | 494753-69-4 | MFCD28124355 | 99.9% | 767.00 g/mol | C42H70O12 | 10 MG
SDP

Catalog No. 5000289429
Encompass_Preferred

Ginsenoside Rk1 is a bioactive triterpenoid saponin derived from heat-processed Panax ginseng, provided as a research-grade analytical standard for biochemical and pharmacological studies.

  • High purity research standard (99.9%).
  • Molecular formula C42H70O12 and molecular weight 767.00 g/mol.
  • Solid, white to light yellow appearance.
  • Storage: solid at -20°C; in solvent, -80°C for 6 months and -20°C for 1 month (protect from light).
  • Available as a 10 mg package suitable for analytical applications.

Catalog No. 50-002-89429 Supplier Medchemexpress LLC Supplier No. HYN251510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ginsenoside Rk1 is a bioactive triterpenoid saponin derived from heat-processed Panax ginseng, provided as a research-grade analytical standard for biochemical and pharmacological studies.

  • High purity research standard (99.9%).
  • Molecular formula C42H70O12 and molecular weight 767.00 g/mol.
  • Solid, white to light yellow appearance.
  • Storage: solid at -20°C; in solvent, -80°C for 6 months and -20°C for 1 month (protect from light).
  • Available as a 10 mg package suitable for analytical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.