Learn More
Medchemexpress LLC HY-N0607 5mg Medchemexpress, Ginsenoside Ro CAS:34367-04-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN06075MG
Encompass_Preferred
Medchemexpress, HY-N0607 5mg Ginsenoside Ro CAS:34367-04-9 Ginsenoside Ro (Polysciasaponin P3; Chikusetsusaponin 5; Chikusetsusaponin V) exhibits a Ca 2+ -antagonistic antiplatelet effect with an IC 50 of 155  μM. Ginsenoside Ro reduces the production of TXA 2 more than it reduces the activities of COX-1 and TXAS. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3638