Learn More
Aviva Systems Biology Corp GIOT-1 Antibody - N-terminal region (ARP36215_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36215_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GIOT-1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human GIOT-1. Alternate names: GIOT1, GIOT-1ApplicationIHC, WB. Species reactivity: Dog, Horse, Human
Catalog No. 50-014-56073