Learn More
American Radiolabeled Chemicals Inc GIP-125I 10UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ARI 013810UC
Encompass
Gastric inhibitory polopeptide-125I (human), 10uCi in buffered solution, 0.2 mCi/ml, S.A. 2200 Ci/mmol, Shipped in Dry Ice
Catalog No. 50-901-013