Learn More
American Radiolabeled Chemicals Inc GIP-125I 25UC
SDP
Catalog No. 50901014
Encompass
Gastric inhibitory polopeptide-125I (human), 25uCi in buffered solution, 0.2 mCi/ml, S.A. 2200 Ci/mmol, Shipped in Dry Ice
Catalog No. 50-901-014 Supplier American Radiolabeled Chemicals Inc Supplier No. ARI 013825UC
This product is regulated by the federal government, and may not be ordered online.
Gastric inhibitory polopeptide-125I (human), 25uCi in buffered solution, 0.2 mCi/ml, S.A. 2200 Ci/mmol, Shipped in Dry Ice