Learn More
Bachem (Pro3)-Gastric Inhibitory Polypeptide (human) Trifluoroacetate
SDP
Supplier:  Bachem 4062141.0500
Encompass_Preferred
(Pro3)-Gastric Inhibitory Polypeptide (human) Trifluoroacetate, H-Tyr-Ala-Pro-Gly-Thr-Phe-Ile-Ser-Asp-Tyr-Ser-Ile-Ala-Met-Asp-Lys-Ile-His-Gln-Gln-Asp-Phe-Val-Asn-Trp-Leu-Leu-Ala-Gln-Lys-Gly-Lys-Lys-Asn-Asp-Trp-Lys-His-Asn-Ile-Thr-Gln-OH, 299898-52-5, C226H338N60O64S, Mr 4951.60, 0.5mg. (Pro3)-GIP (human), (Pro3)-Glucose-Dependent Insulinotropic Polypeptide (human), (Pro3) Gastric Inhibitory Peptide (GIP), human. GIP antagonist.
Catalog No. 50-194-6887