Learn More
Eurofins Discoverx GIP (Gastric Inhibitory Peptide), 500 µg. (92-1078)
Supplier:  Eurofins Discoverx 921078
Encompass
Gastric inhibitory polypeptide (GIP) is a secretin family endocrine peptide hormone produced in the small intestine that induces insulin secretion in a glucose-dependent manner and in response to eating. It also weakly inhibits gastric acid secretion. Acts as an agonist for GIPR. Provided as 0.5 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9618