Learn More
Phoenix Pharmaceuticals Inc GIP (Human) - 100 µg Peptide, ≥ 95% Purity, Stimulates Insulin Secretion
Supplier:  Phoenix Pharmaceuticals Inc 02702
Encompass
GIP (Human) is a high-purity synthetic peptide vital for research in glucose homeostasis and metabolic studies. This 42-amino acid peptide stimulates insulin secretion from pancreatic beta-cells, regulating blood glucose levels. Provided as a lyophilized white powder, it offers stability and ease of handling. Its purity of ≥ 95% and a molecular weight of 4983.6 make it a reliable choice for scientific applications in metabolic research.
Catalog No. NC9048828