Learn More
Medchemexpress LLC GIP Human HEK293 10ug
SDP
Catalog No. 5000179093
Encompass_Preferred
The GIP protein is a member of the glucagon superfamily an important incretin hormone that stimulates pancreatic beta cells to secrete insulin in response to food intake Through its G protein-coupled receptor it activates adenylyl cyclase and other signal transduction pathways GIP Protein Human (HEK293 hFc) is the recombinant human-derived GIP protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-79093 Supplier Medchemexpress LLC Supplier No. HYP74125A10UG
The GIP protein is a member of the glucagon superfamily an important incretin hormone that stimulates pancreatic beta cells to secrete insulin in response to food intake Through its G protein-coupled receptor it activates adenylyl cyclase and other signal transduction pathways GIP Protein Human (HEK293 hFc) is the recombinant human-derived GIP protein expressed by HEK293 with C-hFc labeled tag