Learn More
Medchemexpress LLC HY-P0276A 1mg Medchemexpress, GIP, human (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0276A1MG
Encompass_Preferred
Medchemexpress, HY-P0276A 1mg GIP, human (TFA) CAS: GIP, human TFA is thought to act as an inhibitor of gastric functions. GIP, human TFA, a peptide hormone consisting of 42 amino acids, is a weak inhibitor of gastric acid secretion and a potent stimulator of insulin post meals. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3032