Learn More
Medchemexpress LLC GIP, rat TFA | 99.8% | 5002.58 | 500 UG
SDP
Catalog No. 5000399698
Encompass_Preferred
GIP, rat TFA is a bioactive peptide of rat origin, also known as glucose-dependent insulinotropic polypeptide or gastric inhibitory polypeptide. This 42-amino acid peptide is released by K cells in the duodenum and jejunum in response to food intake. As a member of the insulinotropic hormone peptide family, it stimulates insulin secretion from pancreatic beta cells and appears to promote beta cell proliferation and survival. Recent studies suggest that GIP plays a role in lipid homeostasis and may be involved in the pathogenesis of obesity. It is for research use only.
  • Bioactive peptide of rat origin
  • 42-amino acid peptide
  • Stimulates insulin secretion from pancreatic beta cells
  • Promotes beta cell proliferation and survival
  • Involved in lipid homeostasis and obesity pathogenesis (potential)
  • For research use only
Catalog No. 50-003-99698 Supplier Medchemexpress LLC Supplier No. HYP5390A500UG
GIP, rat TFA is a bioactive peptide of rat origin, also known as glucose-dependent insulinotropic polypeptide or gastric inhibitory polypeptide. This 42-amino acid peptide is released by K cells in the duodenum and jejunum in response to food intake. As a member of the insulinotropic hormone peptide family, it stimulates insulin secretion from pancreatic beta cells and appears to promote beta cell proliferation and survival. Recent studies suggest that GIP plays a role in lipid homeostasis and may be involved in the pathogenesis of obesity. It is for research use only.
  • Bioactive peptide of rat origin
  • 42-amino acid peptide
  • Stimulates insulin secretion from pancreatic beta cells
  • Promotes beta cell proliferation and survival
  • Involved in lipid homeostasis and obesity pathogenesis (potential)
  • For research use only