Learn More
Aviva Systems Biology Corp GIPC2 Antibody - N-terminal region (ARP41271_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41271_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GIPC2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human GIPC2. Alternate names: SEMCAP2, SEMCAP-2ApplicationIHC, WB. Species reactivity: Human, Mouse, Rat
Catalog No. 50-014-57788