Learn More
Aviva Systems Biology Corp GIPC3 Antibody - middle region (ARP41293_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41293_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GIPC3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human GIPC3. Alternate names: C19orf64, DKFZp686J1198, FLJ40925, DFNB15, DFNB72, DFNB95ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Rat
Catalog No. 50-284-924