Learn More
Aviva Systems Biology Corp GIPR Antibody - N-terminal region (ARP63933_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63933_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GIPR. It was validated on Western Blot. . Alternate names: MGC126722, PGQTL2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-122