Learn More
Eurofins Discoverx Ready-to-Assay GIP Glucagon Receptor Frozen Cells, 2 Vials of Cells. (HTS134RTA)
Catalog No. 502005951
Encompass
The Ready-to-Assay GIP Glucagon Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. GIP) based activation of GIPR GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses GIPR alongside exogenous levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of GIPR, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.
Catalog No. 50-200-5951 Supplier Eurofins Discoverx Supplier No. HTS134RTA
The Ready-to-Assay GIP Glucagon Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. GIP) based activation of GIPR GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses GIPR alongside exogenous levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of GIPR, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.