Learn More
Medchemexpress LLC GIPR protein, cynomolgus (P. pastoris, His) | 96.0% | 50UG
SDP
Catalog No. 5000174170
Encompass_Preferred
Recombinant cynomolgus GIPR protein expressed in Pichia pastoris and supplied as a lyophilized fragment with an N-terminal 6xHis tag (accession A0A2K5VD75, residues G26-Q138). Intended for biochemical and biophysical applications, including binding studies, antibody screening, and assay development. Purity ≥90% (96.0% by reducing SDS-PAGE); molecular weight 20-22 kDa.
  • Lyophilized format for long-term storage.
  • His-tag facilitates purification and detection.
  • High purity suitable for biochemical assays.
  • Recommended storage at -20°C; freeze aliquots at -80°C for extended stability.
  • Stable at 4°C for 1 week after reconstitution.
  • Expressed in Pichia pastoris to support eukaryotic processing.
Catalog No. 50-001-74170 Supplier Medchemexpress LLC Supplier No. HYP70280350UG
Recombinant cynomolgus GIPR protein expressed in Pichia pastoris and supplied as a lyophilized fragment with an N-terminal 6xHis tag (accession A0A2K5VD75, residues G26-Q138). Intended for biochemical and biophysical applications, including binding studies, antibody screening, and assay development. Purity ≥90% (96.0% by reducing SDS-PAGE); molecular weight 20-22 kDa.
  • Lyophilized format for long-term storage.
  • His-tag facilitates purification and detection.
  • High purity suitable for biochemical assays.
  • Recommended storage at -20°C; freeze aliquots at -80°C for extended stability.
  • Stable at 4°C for 1 week after reconstitution.
  • Expressed in Pichia pastoris to support eukaryotic processing.