Learn More
Cayman Chemical GIPR EXrAClular DomaInhu 100ug
SDP
Catalog No. 502943453
Encompass_Preferred
Source: Active recombinant human C-terminal His-tagged GIPR extracellular domain expressed in HEK293 cells • Amino acids: 26-138 • MW: 14.42 kDa
Catalog No. 50-294-3453 Supplier Cayman Chemical Supplier No. 42230100 UG
Source: Active recombinant human C-terminal His-tagged GIPR extracellular domain expressed in HEK293 cells • Amino acids: 26-138 • MW: 14.42 kDa