Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GIPR protein, human (HEK293), His-tagged | 10UG
SDP

Catalog No. 5000175969
Encompass_Preferred

Recombinant human GIPR protein (extracellular fragment R22-Q138) expressed in HEK293 and supplied with a C-terminal His tag. Supplied lyophilized from a Tris-NaCl-trehalose buffer, it is intended for receptor-binding and biochemical assays, antibody development, and related research applications. Apparent molecular weight is ~22-28 kDa due to glycosylation.

  • Human GIPR extracellular region (R22-Q138).
  • Expressed in HEK293 cells with a C-terminal His tag.
  • Lyophilized from 20 mM Tris-HCl, 0.5 M NaCl, 6% trehalose, pH 8.0.
  • Apparent molecular weight ~22-28 kDa (glycosylated).
  • Endotoxin <1 EU/μg (LAL method).
  • Store at -20°C; aliquot and freeze at -20°C or -80°C for extended storage.
  • Suitable for receptor-binding and biochemical assays.

Catalog No. 50-001-75969 Supplier Medchemexpress LLC Supplier No. HYP70280010UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human GIPR protein (extracellular fragment R22-Q138) expressed in HEK293 and supplied with a C-terminal His tag. Supplied lyophilized from a Tris-NaCl-trehalose buffer, it is intended for receptor-binding and biochemical assays, antibody development, and related research applications. Apparent molecular weight is ~22-28 kDa due to glycosylation.

  • Human GIPR extracellular region (R22-Q138).
  • Expressed in HEK293 cells with a C-terminal His tag.
  • Lyophilized from 20 mM Tris-HCl, 0.5 M NaCl, 6% trehalose, pH 8.0.
  • Apparent molecular weight ~22-28 kDa (glycosylated).
  • Endotoxin <1 EU/μg (LAL method).
  • Store at -20°C; aliquot and freeze at -20°C or -80°C for extended storage.
  • Suitable for receptor-binding and biochemical assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.