Learn More
Medchemexpress LLC GIPR protein, human (HEK293, His) | 20UG
SDP
Catalog No. 5000175183
Encompass_Preferred
Recombinant human glucose-dependent insulinotropic polypeptide receptor (GIPR) expressed in HEK293 cells and supplied as a His-tagged, lyophilized protein (20 μg). The product is glycosylated, has an apparent molecular weight of ~22-28 kDa, and features low endotoxin levels (<1 EU/μg). It is formulated in Tris-HCl/NaCl/trehalose and is intended for receptor binding, ligand screening, biochemical characterization, and assay development. Store at -20°C and follow reconstitution guidance.
  • Expressed in mammalian cells for native glycosylation
  • His tag facilitates purification and detection
  • Lyophilized format improves shelf stability
  • Low endotoxin suitable for cell-based assays
  • Formulated with trehalose for stabilization on storage
Catalog No. 50-001-75183 Supplier Medchemexpress LLC Supplier No. HYP70280020UG
Recombinant human glucose-dependent insulinotropic polypeptide receptor (GIPR) expressed in HEK293 cells and supplied as a His-tagged, lyophilized protein (20 μg). The product is glycosylated, has an apparent molecular weight of ~22-28 kDa, and features low endotoxin levels (<1 EU/μg). It is formulated in Tris-HCl/NaCl/trehalose and is intended for receptor binding, ligand screening, biochemical characterization, and assay development. Store at -20°C and follow reconstitution guidance.
  • Expressed in mammalian cells for native glycosylation
  • His tag facilitates purification and detection
  • Lyophilized format improves shelf stability
  • Low endotoxin suitable for cell-based assays
  • Formulated with trehalose for stabilization on storage