Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GIPR protein, human (HEK293, His) | 50UG
SDP

Catalog No. 5000174173
Encompass_Preferred

Recombinant human glucose-dependent insulinotropic polypeptide receptor (GIPR) expressed in HEK293 cells, supplied as a His-tagged, lyophilized protein for binding and functional assays. The product covers residues R22-Q138 (Accession P48546-1), is low endotoxin, and includes recommended storage and reconstitution guidance.

  • Recombinant human GIPR (residues R22-Q138) with C-terminal His tag.
  • Expressed in HEK293 mammalian cells for native glycosylation.
  • Lyophilized from 20 mM Tris-HCl, 0.5 M NaCl, 6% trehalose, pH 8.0.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Approximate molecular weight 22-28 kDa due to glycosylation.
  • Functional activity: ED50 = 19.68 ng/mL in binding assay.
  • Storage and handling recommendations included for long-term stability.

Catalog No. 50-001-74173 Supplier Medchemexpress LLC Supplier No. HYP70280050UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human glucose-dependent insulinotropic polypeptide receptor (GIPR) expressed in HEK293 cells, supplied as a His-tagged, lyophilized protein for binding and functional assays. The product covers residues R22-Q138 (Accession P48546-1), is low endotoxin, and includes recommended storage and reconstitution guidance.

  • Recombinant human GIPR (residues R22-Q138) with C-terminal His tag.
  • Expressed in HEK293 mammalian cells for native glycosylation.
  • Lyophilized from 20 mM Tris-HCl, 0.5 M NaCl, 6% trehalose, pH 8.0.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Approximate molecular weight 22-28 kDa due to glycosylation.
  • Functional activity: ED50 = 19.68 ng/mL in binding assay.
  • Storage and handling recommendations included for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.