Learn More
Medchemexpress LLC GIPR protein, human (HEK293, His) | 97.0% | 5UG
SDP
Catalog No. 5000174097
Encompass_Preferred
Recombinant fragment of the human glucose-dependent insulinotropic polypeptide receptor (GIPR) expressed in HEK293 cells and supplied as a His-tagged, lyophilized protein for research applications. It is formulated for stability and provided with storage and reconstitution guidance for biochemical and cell-based assays.
  • Recombinant human GIPR extracellular domain, His-tagged.
  • Expressed in HEK293 cells for mammalian glycosylation.
  • Lyophilized formulation for long-term storage.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • High purity as determined by reducing SDS-PAGE.
  • Recommended reconstitution to ≥100 μg/mL and aliquot storage.
Catalog No. 50-001-74097 Supplier Medchemexpress LLC Supplier No. HYP7028005UG
Add to Cart
Add to Cart
Recombinant fragment of the human glucose-dependent insulinotropic polypeptide receptor (GIPR) expressed in HEK293 cells and supplied as a His-tagged, lyophilized protein for research applications. It is formulated for stability and provided with storage and reconstitution guidance for biochemical and cell-based assays.
  • Recombinant human GIPR extracellular domain, His-tagged.
  • Expressed in HEK293 cells for mammalian glycosylation.
  • Lyophilized formulation for long-term storage.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • High purity as determined by reducing SDS-PAGE.
  • Recommended reconstitution to ≥100 μg/mL and aliquot storage.