Learn More
Medchemexpress LLC GIPR protein, mouse (HEK293, His) | 100UG
SDP
Supplier:  Medchemexpress LLC HYP702801100UG
Encompass_Preferred
Recombinant mouse GIPR protein representing the extracellular domain (E19-Q134), expressed in HEK293 and supplied as a purified, His-tagged protein. Provided as lyophilized material with stabilizing formulations; suitable for biochemical and cell-based assays requiring the receptor extracellular region.
  • Expressed in HEK293 cells for mammalian glycosylation.
  • His tag enables straightforward purification and detection.
  • Extracellular domain sequence corresponds to accession Q0P543-1 (E19-Q134).
  • Glycosylation results in an apparent molecular weight range of ~19-33 kDa.
  • Formulated with trehalose in Tris or PBS for improved stability.
  • Stable at -20°C for long-term storage; short-term stability at 4°C after reconstitution.
  • Suitable for biochemical characterization and cell-based receptor studies.
Catalog No. 50-001-74055