Learn More
Medchemexpress LLC GIPR protein, mouse (HEK293, His-tag) | 50UG

Supplier: Medchemexpress LLC HYP70280150UG
Recombinant extracellular domain of mouse glucose-dependent insulinotropic polypeptide receptor (GIPR) expressed in HEK293 cells and supplied with a C-terminal 10xHis tag. The product is 116 amino acids (E19-Q134), supplied lyophilized with lot-specific formulation; predicted molecular weight is 14.7 kDa and observed as ~19-33 kDa due to glycosylation. Endotoxin level is <1 EU/μg and recommended uses include binding assays and antibody validation.
- Recombinant mouse GIPR extracellular domain, HEK293 expressed.
- C-terminal 10xHis tag for purification and detection.
- 116 amino acids (E19-Q134).
- Predicted MW 14.7 kDa; observed ~19-33 kDa due to glycosylation.
- Lyophilized with lot-specific buffer; COA lists exact formulation.
- Endotoxin level <1 EU/μg.
- Suitable for ELISA, antibody validation, and binding studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.