Learn More
Cayman Chemical Girard's Reagent P, Purity: ≥95%, CAS: 1126-58-5, 25 g
SDP
Catalog No. 502896237
Encompass_Preferred
A cationic hydrazine reagent; has been used as a derivatization reagent to quantify glycans, oxysterols, and 5-methylcytosine derivatives
Catalog No. 50-289-6237 Supplier Cayman Chemical Supplier No. 60154125 G
A cationic hydrazine reagent; has been used as a derivatization reagent to quantify glycans, oxysterols, and 5-methylcytosine derivatives