Learn More
Cayman Chemical GIrards Reagent P-d5 10mg
SDP
Catalog No. 502987402
Encompass_Preferred
An internal standard for the quantification of Girard's reagent P by GC- or LC-MS
Catalog No. 50-298-7402 Supplier Cayman Chemical Supplier No. 2200510 MG
An internal standard for the quantification of Girard's reagent P by GC- or LC-MS