Learn More
Cayman Chemical Girard’s Reagent P-d5, Purity: ≥99% deuterated forms (d1-d5), CAS: 1505505-87-2, 50 mg
SDP
Catalog No. 502987598
Encompass_Preferred
An internal standard for the quantification of Girard's reagent P by GC- or LC-MS
Catalog No. 50-298-7598 Supplier Cayman Chemical Supplier No. 2200550 MG
An internal standard for the quantification of Girard's reagent P by GC- or LC-MS