Learn More
Medchemexpress LLC HY-109176 5mg Medchemexpress, Giredestrant CAS:1953133-47-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1091765MG
Encompass_Preferred
Medchemexpress, HY-109176 5mg Giredestrant CAS:1953133-47-5 Giredestrant (GDC-9545), a non-steroidal estrogen receptor (ER) ligand, is an orally active and selective ER antagonist. Giredestrant potently competes with Estradiol for binding and induces a conformational change within the ER ligand binding domain. Giredestrant has anti-tumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0767