Learn More
TARGETMOL CHEMICALS INC GIRIPLADIB 5MG
SDP
Catalog No. 502580343
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Giripladib (PLA695)(PLX-695) is a cPLA2-specific inhibitor that inhibits the radiation-induced increase of phosphorylated ERK and phosphorylated Akt in endothelial cells. purity: 99%
Catalog No. 50-258-0343 Supplier TARGETMOL CHEMICALS INC Supplier No. T319295MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Giripladib (PLA695)(PLX-695) is a cPLA2-specific inhibitor that inhibits the radiation-induced increase of phosphorylated ERK and phosphorylated Akt in endothelial cells. purity: 99%