Learn More
Medchemexpress LLC Gisadenafil besylate | 334827-98-4 | MFCD18384964 | 99.6% | 677.79 g/mol | C29H39N7O8S2 | 5 MG
SDP
Catalog No. 5000264767
Encompass_Preferred
Gisadenafil besylate (CAS 334827-98-4) is an orally active phosphodiesterase 5 (PDE5) inhibitor used in biochemical and pharmacological research to prevent degradation of cyclic guanosine monophosphate (cGMP). It is employed to study PDE5-mediated signaling and pharmacology in vitro and in vivo, and is provided as a high-purity research reagent.
Catalog No. 50-002-64767 Supplier Medchemexpress LLC Supplier No. HY1086195MG
Gisadenafil besylate (CAS 334827-98-4) is an orally active phosphodiesterase 5 (PDE5) inhibitor used in biochemical and pharmacological research to prevent degradation of cyclic guanosine monophosphate (cGMP). It is employed to study PDE5-mediated signaling and pharmacology in vitro and in vivo, and is provided as a high-purity research reagent.