Learn More
Cayman Chemical GIsadenafIlbesylate 10mg
SDP
Catalog No. 502991177
Encompass_Preferred
A PDE5 inhibitor (IC50 = 1.1 nM); 103-fold selective for PDE5 over PDE6 at 1.1 nM; restores the wild-type response to hypercapnia in the Tat-tg mouse model of HIV-1 neuroinflammation
Catalog No. 50-299-1177 Supplier Cayman Chemical Supplier No. 3147210 MG
A PDE5 inhibitor (IC50 = 1.1 nM); 103-fold selective for PDE5 over PDE6 at 1.1 nM; restores the wild-type response to hypercapnia in the Tat-tg mouse model of HIV-1 neuroinflammation