Learn More
Aviva Systems Biology Corp GIT2 Antibody - C-terminal region (ARP61662_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61662_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GIT2. It was validated on Western Blot. . Alternate names: CAT-2, DKFZp686G01261, KIAA0148, MGC760, CAT2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-287-433