Learn More
Medchemexpress LLC Gitoxin | 4562-36-1 | MFCD00021173 | 95.0% | 780.94 | C41H64O14 | 5 MG
SDP
Catalog No. 5000221986
Encompass_Preferred
Gitoxin is a cardenolide (cardiac) glycoside that inhibits the Na+/K+-ATPase. Used as an analytical standard and research reagent, it commonly appears as a metabolic degradation product of digitoxin and is supplied in small milligram quantities suitable for chromatographic, mass spectrometry, and biochemical assays.
Catalog No. 50-002-21986 Supplier Medchemexpress LLC Supplier No. HY1369335MG
Gitoxin is a cardenolide (cardiac) glycoside that inhibits the Na+/K+-ATPase. Used as an analytical standard and research reagent, it commonly appears as a metabolic degradation product of digitoxin and is supplied in small milligram quantities suitable for chromatographic, mass spectrometry, and biochemical assays.