Learn More
ACROBiosystems ACROBiosystems Rhesus macaque GITR / TNFRSF18 Protein, Fc Tag
Supplier:  ACROBiosystems GIRC52551MG
Encompass_Preferred
Recombinant Protein;1MG;Rhesus macaque GITR, Fc Tag (GIR-C5255) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Glu 161 (Accession # F7DDV8).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 41.3 kDa. As a result of glycosylation, the protein migrates as 48 kDa under reducing (R) condition, and 85 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-195-7407