Learn More
ACROBiosystems ACROBiosystems Biotinylated Human GITR / TNFRSF18 Protein, Fc,Avitag™
Supplier:  ACROBiosystems GIRH82F7200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human GITR, Fc,Avitag (GIR-H82F7) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Glu 161 (Accession # Q9Y5U5-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 43.3 kDa. The protein migrates as 45-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7146