Learn More
ACROBiosystems ACROBiosystems Human GITR / TNFRSF18 Protein, His Tag
Catalog No. 501957145
Encompass_Preferred
Recombinant Protein;1MG;Human GITR, His Tag (GIR-H5228) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Glu 161 (Accession # Q9Y5U5-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.4 kDa. The protein migrates as 22-25 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human GITR Ligand, Fc Tag (Cat. No. GIL-H526a) at 10 μg/mL (100 μL/well) can bind Human GITR, His Tag (Cat. No. GIR-H5228) with a linear range of 1-20 ng/mL (QC tested).
Catalog No. 50-195-7145 Supplier ACROBiosystems Supplier No. GIRH52281MG
Recombinant Protein;1MG;Human GITR, His Tag (GIR-H5228) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Glu 161 (Accession # Q9Y5U5-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.4 kDa. The protein migrates as 22-25 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human GITR Ligand, Fc Tag (Cat. No. GIL-H526a) at 10 μg/mL (100 μL/well) can bind Human GITR, His Tag (Cat. No. GIR-H5228) with a linear range of 1-20 ng/mL (QC tested).