Learn More
ACROBiosystems ACROBiosystems Canine GITR / TNFRSF18 Protein, His Tag
Catalog No. 501956927
Encompass_Preferred
Recombinant Protein;100UG;Canine GITR, His Tag (GIR-C52H6) is expressed from human 293 cells (HEK293). It contains AA Gly 23 - Pro 154 (Accession # D7F619-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.8 kDa. The protein migrates as 20-25 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Canine GITR, His Tag (Cat. No. GIR-C52H6) at 2 μg/mL (100 μL/well) can bind Human GITR Ligand, Fc Tag (Cat. No. GIL-H526a) with a linear range of 2-40 ng/mL (QC tested).
Catalog No. 50-195-6927 Supplier ACROBiosystems Supplier No. GIRC52H6100UG
Recombinant Protein;100UG;Canine GITR, His Tag (GIR-C52H6) is expressed from human 293 cells (HEK293). It contains AA Gly 23 - Pro 154 (Accession # D7F619-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.8 kDa. The protein migrates as 20-25 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Canine GITR, His Tag (Cat. No. GIR-C52H6) at 2 μg/mL (100 μL/well) can bind Human GITR Ligand, Fc Tag (Cat. No. GIL-H526a) with a linear range of 2-40 ng/mL (QC tested).