Learn More
AdipoGen GITR human pAb
SDP
Supplier:  AdipoGen AG25A0017C100
Encompass_Preferred
Polyclonal Antibody. Recognizes human GITR. Does not cross-react with mouse GITR. Source Rat. Applications ELISA, FACS. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. GITR is the receptor for TNFSF18. It is involved in interactions between activated T-lymphocytes and endothelial cells and in the regulation of T-cell receptor-mediated cell death. Mediates NF-kappaB activation via the TRAF2/NIK pathway.
Catalog No. 50-256-3294