Learn More
Biotium GITR / Tnfrsf18 (Mouse)(DTA-1), CF740 conjugate, 0.1mg/mL
Catalog No. 5000624280
Encompass_Preferred
GITR (glucocorticoid-induced TNFR-related gene) is a member of the TNF-receptor superfamily, also known as TNFRSF18. It is expressed at low levels on resting T lymphocytes and at high levels on CD25 CD4 Tregs. The expression of GITR on T cells can be upregulated upon activation. Interaction of GITR with its ligand (GITRL) has been demonstrated to augment T cell activation, proliferation, cytokine production as well as MAPKs and NF-B activation, and abrogate the inhibitory function ofCD25 CD4 Tregs. In vivo activation of GITR causes development of autoimmune diseases and restores the suppressed immune response.
Catalog No. 50-006-24280 Supplier Biotium Supplier No. BNC740976500
GITR (glucocorticoid-induced TNFR-related gene) is a member of the TNF-receptor superfamily, also known as TNFRSF18. It is expressed at low levels on resting T lymphocytes and at high levels on CD25 CD4 Tregs. The expression of GITR on T cells can be upregulated upon activation. Interaction of GITR with its ligand (GITRL) has been demonstrated to augment T cell activation, proliferation, cytokine production as well as MAPKs and NF-B activation, and abrogate the inhibitory function ofCD25 CD4 Tregs. In vivo activation of GITR causes development of autoimmune diseases and restores the suppressed immune response.