Learn More
Genscript Corporation GITR/TNFRSF18Biotin His A
Catalog No. 5000058918
Encompass
GITR (glucocorticoid-induced tumor necrosis factor receptor) also known as AITR and TNFRSF18 is a 40 kDa transmembrane glycoprotein that functions in immune regulation GIRT is a receptor for TNFSF18 Seems to be involved in interactions between activated T-lymphocytes and endothelial cells and in the regulation of T-cell receptor-mediated cell death Mediated NF-kappa-B activation via the TRAF2/NIK pathway
Catalog No. 50-000-58918 Supplier Genscript Corporation Supplier No. Z04474100
GITR (glucocorticoid-induced tumor necrosis factor receptor) also known as AITR and TNFRSF18 is a 40 kDa transmembrane glycoprotein that functions in immune regulation GIRT is a receptor for TNFSF18 Seems to be involved in interactions between activated T-lymphocytes and endothelial cells and in the regulation of T-cell receptor-mediated cell death Mediated NF-kappa-B activation via the TRAF2/NIK pathway