Learn More
ACROBiosystems Recombinant Protein;Human GITR / TNFRSF18 Protein, Fc Tag;HEK293;1MG;GIR,AITR,GITR,TNFRSF18,CD357
Supplier:  ACROBiosystems GIRH52541MG
Encompass_Preferred
Recombinant Protein;Human GITR / TNFRSF18 Protein, Fc Tag;1MG;Human GITR, Fc Tag (GIR-H5254) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Glu 161 (Accession # Q9Y5U5-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 41.0 kDa. The protein migrates as 45-47 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human GITR, Fc Tag (Cat. No. GIR-H5254) at 2 μg/mL (100 μL/well) can bind Biotinylated Human GITR Ligand, Fc Tag, Avi Tag (Cat. No. GIL-H82F8) with a linear range of 3.9-62.5 ng/mL (QC tested).
Catalog No. 50-165-7937