Learn More
Cayman Chemical GITRL EXrAClular DomaInh 100ug
SDP
Catalog No. 502948318
Encompass_Preferred
Source: Recombinant C-terminal mouse IgG2a Fc-tagged human GITRL expressed in HEK293 cells • Amino acids: 72-199 • MW: 40.9 kDa
Catalog No. 50-294-8318 Supplier Cayman Chemical Supplier No. 32018100 UG
Source: Recombinant C-terminal mouse IgG2a Fc-tagged human GITRL expressed in HEK293 cells • Amino acids: 72-199 • MW: 40.9 kDa