Learn More
AdipoGen GITRL human pAb
SDP
Supplier:  AdipoGen AG25A0023C100
Encompass_Preferred
Polyclonal Antibody. Recognizes human GITRL. Detects a band of ~15-17kDa by Western blot. Does not cross-react with mouse GITRL. Source Rabbit. Applications ELISA, IHC, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. GITRL is expressed on dendritic cells DC, monocytes, macrophages, B cells, activated T cells, endothelial cells, osteoclasts and various healthy non-lymphoid tissues e.g. testis, .. GITRL is constitutively expressed and released as soluble form by solid tumors and various hematopoietic malignancies. GITRL causes differentiation of osteoclasts, activation of macrophages, but also alteration of carcinoma and leukemia cells and influences apoptosis. Binding to GITR is important in regulating T-cell proliferation and TCR-mediated apoptosis. GITRL is implicated in development of autoimmune diseases and in the immune response against infectious pathogens and tumors.
Catalog No. 50-256-3289