Learn More
AdipoGen GITRL Soluble mouse rec
SDP
Catalog No. 502563064
Encompass_Preferred
Protein. The extracellular domain of mouse GITRL aa 45-173 is fused at the N-terminus to a FLAG -tag. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in 10mM glycine, pH 10.0. Binds to mouse GITR. Purity >90% SDS-PAGE. GITRL Glucocorticoid-induced TNF receptor ligand is expressed on dendritic cells DC, monocytes, macrophages, B cells, activated T cells, endothelial cells, osteoclasts and various healthy non-lymphoid tissues e.g. testis. GITRL is constitutively expressed and released as soluble form by solid tumors and various hematopoietic malignancies. GITRL causes differentiation of osteoclasts, activation of macrophages, but also alteration of carcinoma and leukemia cells and influences apoptosis. Binding to GITR is important in regulating T cell proliferation and TCR-mediated apoptosis.
Catalog No. 50-256-3064 Supplier AdipoGen Supplier No. AG40A0008C050
Protein. The extracellular domain of mouse GITRL aa 45-173 is fused at the N-terminus to a FLAG -tag. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in 10mM glycine, pH 10.0. Binds to mouse GITR. Purity >90% SDS-PAGE. GITRL Glucocorticoid-induced TNF receptor ligand is expressed on dendritic cells DC, monocytes, macrophages, B cells, activated T cells, endothelial cells, osteoclasts and various healthy non-lymphoid tissues e.g. testis. GITRL is constitutively expressed and released as soluble form by solid tumors and various hematopoietic malignancies. GITRL causes differentiation of osteoclasts, activation of macrophages, but also alteration of carcinoma and leukemia cells and influences apoptosis. Binding to GITR is important in regulating T cell proliferation and TCR-mediated apoptosis.