Learn More
ACROBiosystems ACROBiosystems Human GITR Ligand / TNFSF18 Protein, His Tag
Supplier:  ACROBiosystems GILH52491MG
Encompass_Preferred
Recombinant Protein;1MG;Human GITR Ligand, His Tag (GIL-H5249) is expressed from human 293 cells (HEK293). It contains AA Gln 50 - Ser 177 (Accession # AAH69319.1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 20.5 kDa. The protein migrates as 23 kDa and 26 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human GITR Ligand, His Tag (Cat. No. GIL-H5249) at 2 μg/mL (100 μL/well) can bind Human GITR, Fc Tag (Cat. No. GIR-H5254) with a linear range of 0.1-4 ng/mL (QC tested).;Immobilized Human GITR Ligand, His Tag (Cat. No. GIL-H5249) at 2 μg/mL (100 μL/well) can bind Human GITR, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. GIR-H525a) with a linear range of 0.2-8 ng/mL (Routinely tested).
Catalog No. 50-195-7101