Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human GITR Ligand / TNFSF18 Protein, Fc Tag, Avi Tag (Avitag™);HEK293;25UG;GIL,TNFSF18,AITRL,GITRL,TL6,hGITRL,TNFSF-18,GITR Ligand
Catalog No. 501657934
Encompass_Preferred
Recombinant Protein;Biotinylated Human GITR Ligand / TNFSF18 Protein, Fc Tag, Avi Tag (Avitag™);25UG;Biotinylated Human GITR Ligand, Fc Tag, Avi Tag (GIL-H82F8) is expressed from human 293 cells (HEK293). It contains AA Gln 50 - Ser 177 (Accession # AAH69319.1).;This protein carries an Avi tag (Avitag™) at the N-terminus, followed by a human IgG1 Fc tag. The protein has a calculated MW of 43.5 kDa. The protein migrates as 45-53 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human GITR, Fc Tag (Cat. No. GIR-H5254) at 2 μg/mL (100 μL/well) can bind Biotinylated Human GITR Ligand, Fc Tag, Avi Tag (Cat. No. GIL-H82F8) with a linear range of 3.9-62.5 ng/mL (QC tested).
Catalog No. 50-165-7934 Supplier ACROBiosystems Supplier No. GILH82F825UG
Recombinant Protein;Biotinylated Human GITR Ligand / TNFSF18 Protein, Fc Tag, Avi Tag (Avitag™);25UG;Biotinylated Human GITR Ligand, Fc Tag, Avi Tag (GIL-H82F8) is expressed from human 293 cells (HEK293). It contains AA Gln 50 - Ser 177 (Accession # AAH69319.1).;This protein carries an Avi tag (Avitag™) at the N-terminus, followed by a human IgG1 Fc tag. The protein has a calculated MW of 43.5 kDa. The protein migrates as 45-53 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human GITR, Fc Tag (Cat. No. GIR-H5254) at 2 μg/mL (100 μL/well) can bind Biotinylated Human GITR Ligand, Fc Tag, Avi Tag (Cat. No. GIL-H82F8) with a linear range of 3.9-62.5 ng/mL (QC tested).