Learn More
ACROBiosystems Recombinant Protein;Human GITR Ligand / TNFSF18 Protein, Fc Tag;HEK293;50UG;GIL,TNFSF18,AITRL,GITRL,TL6,hGITRL,TNFSF-18,GITR Ligand
Supplier:  ACROBiosystems GILH526A50UG
Encompass_Preferred
Recombinant Protein;Human GITR Ligand / TNFSF18 Protein, Fc Tag;50UG;Human GITR Ligand, Fc Tag (GIL-H526a) is expressed from human 293 cells (HEK293). It contains AA Gln 50 - Ser 177 (Accession # AAH69319.1).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 41.5 kDa. As a result of glycosylation, the protein migrates as 46 kDa under reducing (R) condition, and 90 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human GITR, His Tag (Cat. No. GIR-H5228) at 2 μg/mL (100 μl/well) can bind Human GITR Ligand, Fc Tag (Cat. No. GIL-H526a ) with a linear range of 1-16 ng/mL (QC tested).
Catalog No. 50-165-8342