Learn More
Medchemexpress LLC HY-14842B 5mg Medchemexpress, Givinostat (hydrochloride monohydrate) CAS:732302-99-7 Purity:>98%
SDP
Catalog No. 501969918
Encompass_Preferred
Medchemexpress, HY-14842B 5mg Givinostat (HCl monohydrate) CAS:732302-99-7 Givinostat hydrochloride monohydrate (ITF-2357 hydrochloride monohydrate) is a HDAC inhibitor with an IC50 of 198 and 157 nM for HDAC1 and HDAC3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9918 Supplier Medchemexpress LLC Supplier No. HY14842B5MG
Medchemexpress, HY-14842B 5mg Givinostat (HCl monohydrate) CAS:732302-99-7 Givinostat hydrochloride monohydrate (ITF-2357 hydrochloride monohydrate) is a HDAC inhibitor with an IC50 of 198 and 157 nM for HDAC1 and HDAC3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.