Learn More
TARGETMOL CHEMICALS INC GIVINOSTAT HYDROCHLORIDE 10MG
SDP
Catalog No. 502576545
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Givinostat hydrochloride monohydrate (ITF2357) is an HDAC inhibitor. purity: 98%
Catalog No. 50-257-6545 Supplier TARGETMOL CHEMICALS INC Supplier No. T627910MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Givinostat hydrochloride monohydrate (ITF2357) is an HDAC inhibitor. purity: 98%